MilliporeSigma
  • Disruption of Streptococcus mutans and Candida albicans synergy by a commensal streptococcus.
Disruption of Streptococcus mutans and Candida albicans synergy by a commensal streptococcus.
Scientific reports (2020-11-14)
Joshua T Huffines, Jessica A Scoffield
ABSTRACT
Polymicrobial interactions in dental plaque play a significant role in dysbiosis and homeostasis in the oral cavity. In early childhood caries, Streptococcus mutans and Candida albicans are often co-isolated from carious lesions and associated with increased disease severity. Studies have demonstrated that metabolic and glucan-dependent synergism between C. albicans and S. mutans contribute to enhanced pathogenesis. However, it is unclear how oral commensals influence pathogen synergy. Streptococcus parasanguinis, a hydrogen peroxide (H2O2) producing oral commensal, has antimicrobial activity against S. mutans. In this study, we utilized a three species biofilm model to understand the impact of S. parasanguinis on S. mutans and C. albicans synergy. We report that S. parasanguinis disrupts S. mutans and C. albicans biofilm synergy in a contact and H2O2-independent manner. Further, metabolomics analysis revealed a S. parasanguinis-driven alteration in sugar metabolism that restricts biofilm development by S. mutans. Moreover, S. parasanguinis inhibits S. mutans glucosyltransferase (GtfB) activity, which is important for glucan matrix development and GtfB-mediated binding to C. albicans mannan. Taken together, our study describes a new antimicrobial role for S. parasanguinis and highlights how this abundant oral commensal may be utilized to attenuate pathogen synergism.