MilliporeSigma
Small-molecule regulation of zebrafish gene expression.
Nature chemical biology (2007-01-24)
Hanife Esengil, Vicky Chang, John K Mich, James K Chen
ABSTRACT
The zebrafish has emerged as a versatile model organism for biomedical research, yet its potential has been limited by a lack of conditional reverse-genetic tools. Here we report a chemically inducible gene expression technology that has orthogonality to vertebrate signaling processes, high induction levels, and rapid kinetics. Coupled with tissue-specific promoters, this system provides multidimensional control of gene expression and will enable new models of human disorders and diseases.
Sigma-Aldrich
Ponasterone A, ≥65%