MilliporeSigma
The lncRNA H19 alleviates muscular dystrophy by stabilizing dystrophin.
Nature cell biology (2020-10-28)
Yaohua Zhang, Yajuan Li, Qingsong Hu, Yutao Xi, Zhen Xing, Zhao Zhang, Lisa Huang, Jianbo Wu, Ke Liang, Tina K Nguyen, Sergey D Egranov, Chengcao Sun, Zilong Zhao, David H Hawke, Jin Li, Deqiang Sun, Jean J Kim, Ping Zhang, Jie Cheng, Abid Farida, Mien-Chie Hung, Leng Han, Radbod Darabi, Chunru Lin, Liuqing Yang
ABSTRACT
Dystrophin proteomic regulation in muscular dystrophies (MDs) remains unclear. We report that a long noncoding RNA (lncRNA), H19, associates with dystrophin and inhibits E3-ligase-dependent polyubiquitination at Lys 3584 (referred to as Ub-DMD) and its subsequent protein degradation. In-frame deletions in BMD and a DMD non-silent mutation (C3340Y) resulted in defects in the ability of the protein to interact with H19, which caused elevated Ub-DMD levels and dystrophin degradation. Dmd C3333Y mice exhibited progressive MD, elevated serum creatine kinase, heart dilation, blood vessel irregularity and respiratory failure with concurrently reduced dystrophin and increased Ub-DMD status. H19 RNA oligonucleotides conjugated with agrin (AGR-H19) and nifenazone competed with or inhibited TRIM63. Dmd C3333Y animals, induced-pluripotent-stem-cell-derived skeletal muscle cells from patients with Becker MD and mdx mice subjected to exon skipping exhibited inhibited dystrophin degradation, preserved skeletal and cardiac muscle histology, and improved strength and heart function following AGR-H19 or nifenazone treatment. Our study paves the way for meaningful targeted therapeutics for Becker MD and for certain patients with Duchenne MD.
Sigma-Aldrich
MISSION® esiRNA, targeting human TRIM63