MilliporeSigma
  • Arginine methylation-dependent LSD1 stability promotes invasion and metastasis of breast cancer.
Arginine methylation-dependent LSD1 stability promotes invasion and metastasis of breast cancer.
EMBO reports (2019-12-14)
Jiwei Liu, Jingxin Feng, Lili Li, Luyao Lin, Jiafei Ji, Cong Lin, Lingxia Liu, Na Zhang, Dandan Duan, Zhongwei Li, Baiqu Huang, Yu Zhang, Jun Lu
ABSTRACT
Histone lysine demethylase 1 (LSD1), the first identified histone demethylase, is overexpressed in multiple tumor types, including breast cancer. However, the mechanisms that cause LSD1 dysregulation in breast cancer remain largely unclear. Here, we report that protein arginine methyltransferase 4 (PRMT4 or CARM1) dimethylates LSD1 at R838, which promotes the binding of the deubiquitinase USP7, resulting in the deubiquitination and stabilization of LSD1. Moreover, CARM1- and USP7-dependent LSD1 stabilization plays a key role in repressing E-cadherin and activating vimentin transcription through promoter H3K4me2 and H3K9me2 demethylation, respectively, which promotes invasion and metastasis of breast cancer cells. Consistently, LSD1 arginine methylation levels correlate with tumor grade in human malignant breast carcinoma samples. Our findings unveil a unique mechanism controlling LSD1 stability by arginine methylation, also highlighting the role of the CARM1-USP7-LSD1 axis in breast cancer progression.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-LSD1 (AOF2) (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Cholera Toxin A Subunit from Vibrio cholerae, lyophilized powder
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
Anti-LSD1/BHC110 Antibody, ascites fluid, Upstate®
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-dimethyl-Arginine Antibody, asymmetric (ASYM24), serum, Upstate®
Sigma-Aldrich
Anti-CARM1 Antibody, from rabbit