MilliporeSigma
  • Nanoparticles of chosen noble metals as reactive oxygen species scavengers.
Nanoparticles of chosen noble metals as reactive oxygen species scavengers.
Nanotechnology (2020-10-16)
Polina Ivanova, Konrad Dzięgielewski, Marcin Drozd, Sandra Skorupska, Ilona Grabowska-Jadach, Mariusz Pietrzak
ABSTRACT
Reactive oxygen species (ROS) play an important role in various physiological processes of living organisms. However, their increased concentration is usually considered as a threat for our health. Plants, invertebrates, and vertebrates including humans have various enzymatic and non-enzymatic defence systems against ROS. Unfortunately, both bad condition of surrounding environment and unhealthy lifestyle can interfere with an activity of enzymes responsible for a regulation of ROS levels. Therefore, it is important to look for alternative ROS scavengers, which could be administrated to chosen tissues to prevent pathological processes such as distortion of DNA or RNA structures and oxidation of proteins and lipids. One of the most recently proposed solutions is the application of nanozymes, which could mimic the activity of essential enzymes and prevent excessive activity of ROS. In this work, nanoparticles of Au, Pt, Pd, Ru and Rh were synthesized and studied in this regard. Peroxidase-, catalase (CAT)- and superoxide dismutase (SOD)-like activity of obtained nanoparticles were tested and compared using different methods. The influence of bovine and human albumins on CAT- and peroxidase-like activity was examined. Moreover, in the case of CAT-like activity, an influence of pH and temperature was examined and compared. Determination of SOD-like activity using the methods described for the examination of the activity of native enzyme was not fully successful. Moreover, cytotoxicity of chosen nanoparticles was studied on both regular and tumor cells.
Sigma-Aldrich
Chloroplatinic acid solution, 8 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Rhodium(III) chloride, 98%
Pricing and availability is not currently available.