MilliporeSigma
  • MDM2 and CDK4 immunostainings are useful adjuncts in diagnosing well-differentiated and dedifferentiated liposarcoma subtypes: a comparative analysis of 559 soft tissue neoplasms with genetic data.
MDM2 and CDK4 immunostainings are useful adjuncts in diagnosing well-differentiated and dedifferentiated liposarcoma subtypes: a comparative analysis of 559 soft tissue neoplasms with genetic data.
The American journal of surgical pathology (2005-09-15)
Matthieu Bui Nguyen Binh, Xavier Sastre-Garau, Louis Guillou, Gonzague de Pinieux, Philippe Terrier, Réal Lagacé, Alain Aurias, Isabelle Hostein, Jean Michel Coindre
ABSTRACT
Atypical lipomatous tumor/well-differentiated liposarcoma (ALT-WDLPS) and dedifferentiated liposarcoma (DDLPS) may be difficult to distinguish from benign adipose tumors and from poorly differentiated sarcomas, respectively. Genetically, they are characterized by amplification of MDM2 and CDK4 genes on chromosome 12q13-15. We examined a series of 559 soft tissue tumors (44 ALT-WDLPS, 61 DDLPS, 49 benign adipose tumors, and 405 non-ALT-WDLPS/DDLPS sarcomas) for MDM2 and CDK4 expression using immunohistochemistry. MDM2 and CDK4 immunoexpressions were compared with gene amplification status (as assessed by quantitative PCR and/or comparative genomic hybridization) in 241 neoplasms. Most ALT-WDLPS/DDLPS expressed MDM2 (97%) and CDK4 (92%) as opposed to few benign adipose tumors (MDM2, 5%; CDK4, 2%) and a limited number of non-ALT-WDLSP/DDLPS sarcomas (MDM2, 19%; CDK4, 6%). The sensitivity and specificity of MDM2 and CDK4 immunostainings in identifying ALT-WDLPS/DDLPS among other soft tissue tumors were 97% and 92%, and 83% and 95%, respectively. MDM2 and CDK4 immunostainings were particularly useful to separate ALT-WDLPS from the large group of differentiated adipose tumors, and to distinguish DDLPS from poorly differentiated sarcomas. A strong correlation was observed between MDM2 and CDK4 stainings and gene amplification status. In conclusion, MDM2 and CDK4 immunostainings, which correlate with gene amplification, are helpful adjuncts to differentiate ALT-WDLPS from benign adipose tumors and to separate DDLPS from poorly differentiated sarcomas.
Sigma-Aldrich
CDK4 (DCS-31) Mouse Monoclonal Antibody
Sigma-Aldrich
MDM2 (IF2) Mouse Monoclonal Antibody