MilliporeSigma
Fibulin-5 interacts with fibrillin-1 molecules and microfibrils.
The Biochemical journal (2005-03-26)
Lyle J Freeman, Amanda Lomas, Nigel Hodson, Michael J Sherratt, Kieran T Mellody, Anthony S Weiss, Adrian Shuttleworth, Cay M Kielty
ABSTRACT
Fibulin-5 plays an important role in elastic fibre formation in vivo. We have investigated the molecular interactions between fibulin-5 and components of fibrillin-rich microfibrils which form a template for elastin. Fibulin-5 interacted in a dose-dependent manner with a fibrillin-1 N-terminal sequence and with tropoelastin, but not with MAGP-1 (microfibril-associated glycoprotein-1) or decorin. Fibulin-5 did not inhibit interactions between fibrillin-1 N- and C-terminal fragments, or fibrillin-1 interactions with tropoelastin. Fibulin-5 may provide a link between tropoelastin and microfibrils in the pericellular space during elastic fibre assembly.
Product Description
Millipore
ReBlot Plus Mild Antibody Stripping Solution, 10x