MilliporeSigma
  • Swainsonine protects H9c2 cells against lipopolysaccharide-induced apoptosis and inflammatory injury via down-regulating miR-429.
Swainsonine protects H9c2 cells against lipopolysaccharide-induced apoptosis and inflammatory injury via down-regulating miR-429.
Cell cycle (Georgetown, Tex.) (2019-12-27)
Hongyan Lv, Su Zhang, Xiaohong Hao
ABSTRACT
Pediatric myocarditis (PM) is usually related to myocardial dysfunction. Generally, 30% of PM patients will die or undergo heart transplantation. Swainsonine (SW) is a natural alkaloid and an anti-cancer substance. Our goal was to determine the roles of SW in PM in current study. H9c2 cells were pre-treated by lipopolysaccharide (LPS). Viability and apoptosis were evaluated utilizing CCK-8 assay and flow cytometry. Inflammatory cytokines' mRNA expression and production were assessed by western blot and ELISA. Western blot was utilized to distinguish apoptosis and immune-related factors expression. Sequentially, the abovementioned parameters were reassessed when miR-429 was overexpressed. LPS declined viability as well as raised apoptosis and inflammatory injury in H9c2 cells. SW alleviated apoptosis and inflammatory injury induced by LPS. MiR-429 expression was elevated by LPS and suppressed by SW. SW-induced the increasing of viability and the reduction of inflammatory injury were reversed by overexpression of miR-429. Eventually, SW inhibited p38MAPK/NF-κB pathway which activated by LPS via overexpressing miR-429. SW exerted its anti-apoptosis and anti-inflammatory function in LPS-treated H9c2 cells through p38MAPK/NF-κB pathway and down-regulation of miR-429.
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody