MilliporeSigma
  • Retracted: Allopregnanolone restores the tyrosine hydroxylase-positive neurons and motor performance in a 6-OHDA-injected mouse model.
Retracted: Allopregnanolone restores the tyrosine hydroxylase-positive neurons and motor performance in a 6-OHDA-injected mouse model.
CNS neuroscience & therapeutics (2020-07-01)
Zhi-Chi Chen, Tong-Tong Wang, Wei Bian, Xin Ye, Meng-Yi Li, Juan-Juan Du, Peng Zhou, Huai-Rui Cui, Yu-Qiang Ding, Yan-Hua Ren, Shuang-Shuang Qi, Yuan-Yuan Yuan, Min Liao, Chen-You Sun
ABSTRACT
It has been reported that allopregnanolone (APα) promotes the neurogenesis of the neural progenitor cells (NPCs) in the subventricular zone (SVZ) and prevents the decrease of dopaminergic neurons in 6-hydroxydopamine (6-OHDA)-treated mice by binding to γ-aminobutyric acid A receptor (GABAAR) and then opening voltage-gated L-type Ca2+ channel, but the underlying mechanisms remain elusive. The aim of this study was to explore the possible involvement of GABAAR and calcium/calmodulin-dependent protein kinase II delta 3 (CaMKIIδ3) in this process. 6-OHDA-treated mice and primary cultured midbrain cells were administrated with APα and GABAAR antagonist bicuculline (Bic), and the proliferation and differentiation of NPCs, the tyrosine hydroxylase (TH)-positive neurons and their fibers, the expression levels of CaMKIIδ3 and brain-derived neurotrophic factor (BDNF), and motor functions were measured using ELISA, immunohistochemical staining, real-time RT-PCR, Western blot, and behavioral test. Allopregnanolone significantly promoted the phosphorylation of cytoplasmic CaMKIIδ3 and its nuclear translocation by binding to GABAAR, which, in turn, increased the expression levels of BDNF. This may account for the findings that the exogenous APα enhanced the proliferation and differentiation of NPCs, and ameliorated the nigrostriatal system and behavioral performance in 6-OHDA-treated mice. Allopregnanolone may directly activate GABAAR, which, in turn, enhance the proliferation and differentiation of NPCs via upregulating the expression levels of CaMKIIδ3, and finally contribute to the restoration of dopaminergic neurons in 6-OHDA-treated mice.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-BDNF Antibody, clone 8G8.2, clone 8G8.2, from mouse
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-NeuN Antibody, serum, from guinea pig