MilliporeSigma
  • Synthesis and in vitro antimicrobial activity of new 2-[p-substituted-benzyl]-5-[substituted-carbonylamino]benzoxazoles.
Synthesis and in vitro antimicrobial activity of new 2-[p-substituted-benzyl]-5-[substituted-carbonylamino]benzoxazoles.
European journal of medicinal chemistry (2007-03-06)
Betul Tekiner-Gulbas, Ozlem Temiz-Arpaci, Ilkay Yildiz, Nurten Altanlar
ABSTRACT
Some new 2-(benzyl/p-chlorobenzyl)-5-[(substituted-thienyl/phenyl/phenylthiomethyl/benzyl)carbonylamino]benzoxazole derivatives have been synthesized by reacting 5-amino-2-(benzyl/p-chlorobenzyl)benzoxazoles with appropriate carboxylic acid chlorides. The structures of the synthesized compounds were confirmed by IR, (1)H NMR and MASS spectral data. In vitro antimicrobial activities of the compounds were investigated using twofold serial dilution technique against different two Gram-positive, two Gram-negative bacteria and three Candida spp. in comparison with standard drugs. Microbiological results indicated that the newly synthesized 2-(benzyl/p-chlorobenzyl)-5-[(substituted-thienyl/phenyl/phenylthiomethyl/benzyl)carbonylamino]benzoxazole derivatives (3-12) possessed a broad spectrum of activity having MIC values of 6.25-100 microg/ml against the tested microorganisms. Especially, with an MIC value of 6.25 microg/ml, 2-(p-chlorophenyl)-5-[(2,5-dimethylphenyl)carbonylamino]benzoxazole 4 displayed the same activity against Candida albicans as the standard drug clotrimazole.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Clotrimazole
Supelco
Clotrimazole, VETRANAL®, analytical standard