MilliporeSigma
  • Preparation and characterization of a monoclonal antibody with high affinity for soluble Abeta oligomers.
Preparation and characterization of a monoclonal antibody with high affinity for soluble Abeta oligomers.
Hybridoma (2005) (2009-10-28)
Zhang Ying, Wang Xin, He Jin-Sheng, Bao Fu-Xiang, Sun Wei-Min, Dai Xin-Xian, Wang Xiao-Bo, Li Yi-Qin, Zhen Xian-Xian, Hu Hong-Gang, Peng Xiang-Lei, Zheng Yan-Peng, Hou Ling-Ling, Hong Tao
ABSTRACT
Amyloid beta-protein (Abeta) has been causally implicated in the neurodegenerative processes that accompany Alzheimer's disease. Soluble oligomers of the Abeta(1-42) fragment are thought to be significantly more neurotoxic than higher molecular weight aggregates. We report the isolation and characterization of a mouse monoclonal antibody (MAb) directed against soluble Abeta(1-42) oligomers. Synthetic Abeta(1-42) oligomers were assembled in vitro; these were used to immunize mice, and hybridomas were isolated following myeloma fusion of splenocytes from immunized animals. Screening for reactivity against Abeta(1-42) resulted in the identification of MAb A8 with high affinity for soluble oligomers. The isotype of A8 was found to be IgG(2b). Experiments using sub-peptides of Abeta(1-42) revealed that the epitope identified by A8 lies within the 1-6 region of Abeta. The antibody displays high affinity for soluble Abeta(1-42) oligomers in the molecular weight range of 16.5-25 kDa, and detected target antigen in brain sections from senescence-accelerated SAMP 8 mice. The sensitivity and optimal titers for the detection of soluble Abeta(1-42) oligomers were determined to be 0.625 microg/mL in indirect ELISA, and 1:10(6), 1:4000, and 1:150 for ELISA, Western blotting, and immunohistochemistry, respectively. The A8 antibody specific for soluble Abeta(1-42) oligomers will provide a valuable tool for Alzheimer's disease research.
Sigma-Aldrich
Mouse Monoclonal Antibody Isotyping Reagents, sufficient for 1000 tests (clones) (by ELISA), sufficient for 40 tests (clones) (by immunodiffusion, ODD)