MilliporeSigma
  • Probable role for major facilitator superfamily domain containing 6 (MFSD6) in the brain during variable energy consumption.
Probable role for major facilitator superfamily domain containing 6 (MFSD6) in the brain during variable energy consumption.
The International journal of neuroscience (2020-01-08)
Sonchita Bagchi, Emelie Perland, Kimia Hosseini, Johanna Lundgren, Noura Al-Walai, Sania Kheder, Robert Fredriksson
ABSTRACT
Purpose: The major facilitator superfamily (MFS) is known as the largest and most diverse superfamily containing human transporters, and these transporters are essential as they sustain the homeostasis within cellular compartments by moving substances over lipid membranes.Methods: We have identified a novel MFS protein, named Major facilitator superfamily domain containing 6 (MFSD6), and confirmed that it is phylogenetically related to the human Solute Carrier (SLC) transporter family. A homology model of MFSD6 revealed 12 predicted transmembrane segments (TMS) with the classical MFS fold between TMS 6 and 7.Results: Immunohistological analyses showed specific MFSD6 staining in neurons of wildtype mouse brain tissue, but no expression in astrocytes. Furthermore, we explored expression and probable function(s) of MFSD6 in relation to its phylogenetically related proteins, major facilitator superfamily domain containing 8 (MFSD8) and 10 (MFSD10), which is of interest as both these proteins are involved in diseases.Conclusions: We showed that expression levels of Mfsd6 and Mfsd10 were decreased with elevated or depleted energy consumption, while that of Mfsd8 remained unaffected.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Milli-Mark® Pan Neuronal Marker, Milli-Mark Pan Neuronal Marker is an antibody targeting the Pan Neuronal Marker protein, validated for use in ICC, IHC, IF & IHC.
Sigma-Aldrich
Anti-MFSD6 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MFSD8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse