MilliporeSigma
  • 1-Acylthiosemicarbazides, 1,2,4-triazole-5(4H)-thiones, 1,3,4-thiadiazoles and hydrazones containing 5-methyl-2-benzoxazolinones: synthesis, analgesic-anti-inflammatory and antimicrobial activities.
1-Acylthiosemicarbazides, 1,2,4-triazole-5(4H)-thiones, 1,3,4-thiadiazoles and hydrazones containing 5-methyl-2-benzoxazolinones: synthesis, analgesic-anti-inflammatory and antimicrobial activities.
Bioorganic & medicinal chemistry (2007-06-26)
Umut Salgin-Gökşen, Nesrin Gökhan-Kelekçi, Ozgür Göktaş, Yavuz Köysal, Ekrem Kiliç, Samil Işik, Göknur Aktay, Meral Ozalp
ABSTRACT
Acetic acid hydrazide containing 5-methyl-2-benzoxazolinone (4) was synthesized by the condensation of 2-(5-methyl-2-benzoxazolinone-3-yl)acetate with hydrazine hydrate. Thiosemicarbazide derivatives (5a-5d) were afforded by the reaction of corresponding compound 4 with substituted isothiocyanates. The cyclization of compounds 5a-5d in the presence of triethylamine resulted in the formation of compounds 6a-6d containing 1,2,4-triazole ring. On the other hand, the treatment of compounds 5a-5d with orthophosphoric acid caused the conversion of side chain of compounds 5a-5d into 1,3,4-thiadiazole ring: thus, compounds 7a-7c were obtained. The treatment of compound 4 with aromatic aldehydes resulted in the formation of arylidene hydrazides as cis-trans conformers (8a-8e). The structures of the compounds were elucidated by spectral and elemental analysis. While most compounds were exhibiting high activity in the analgesic-anti-inflammatory field, most of them were found to be inactive against bacteria and fungi.
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
1H-1,2,4-Triazole-3-thiol, 97%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%