MilliporeSigma
  • An aberrant SREBP-dependent lipogenic program promotes metastatic prostate cancer.
An aberrant SREBP-dependent lipogenic program promotes metastatic prostate cancer.
Nature genetics (2018-01-18)
Ming Chen, Jiangwen Zhang, Katia Sampieri, John G Clohessy, Lourdes Mendez, Enrique Gonzalez-Billalabeitia, Xue-Song Liu, Yu-Ru Lee, Jacqueline Fung, Jesse M Katon, Archita Venugopal Menon, Kaitlyn A Webster, Christopher Ng, Maria Dilia Palumbieri, Moussa S Diolombi, Susanne B Breitkopf, Julie Teruya-Feldstein, Sabina Signoretti, Roderick T Bronson, John M Asara, Mireia Castillo-Martin, Carlos Cordon-Cardo, Pier Paolo Pandolfi
ABSTRACT
Lipids, either endogenously synthesized or exogenous, have been linked to human cancer. Here we found that PML is frequently co-deleted with PTEN in metastatic human prostate cancer (CaP). We demonstrated that conditional inactivation of Pml in the mouse prostate morphs indolent Pten-null tumors into lethal metastatic disease. We identified MAPK reactivation, subsequent hyperactivation of an aberrant SREBP prometastatic lipogenic program, and a distinctive lipidomic profile as key characteristic features of metastatic Pml and Pten double-null CaP. Furthermore, targeting SREBP in vivo by fatostatin blocked both tumor growth and distant metastasis. Importantly, a high-fat diet (HFD) induced lipid accumulation in prostate tumors and was sufficient to drive metastasis in a nonmetastatic Pten-null mouse model of CaP, and an SREBP signature was highly enriched in metastatic human CaP. Thus, our findings uncover a prometastatic lipogenic program and lend direct genetic and experimental support to the notion that a Western HFD can promote metastasis.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid