MilliporeSigma
  • All-trans retinoic acid controls differentiation, proliferation, and lipolysis in isolated subcutaneous adipocytes from peripartal Holstein cows.
All-trans retinoic acid controls differentiation, proliferation, and lipolysis in isolated subcutaneous adipocytes from peripartal Holstein cows.
Journal of dairy science (2021-02-09)
Qiushi Xu, Yunhui Fan, Juan J Loor, Yusheng Liang, Xudong Sun, Hongdou Jia, Chenxu Zhao, Chuang Xu
ABSTRACT
Preadipocyte proliferation and differentiation are critical for normal adipose tissue development, including achieving a mature phenotype, characterized by its ability to accumulate triacylglycerol and release fatty acids. In nonruminants, it is well known that all-trans retinoic acid (ATRA), the most-active form of vitamin A, helps regulate proliferation, differentiation, and apoptosis in several types of cells including adipocytes. The purpose of this study was to evaluate the effect of ATRA on proliferation, apoptosis, differentiation, and lipolysis of primary bovine adipocytes isolated from subcutaneous adipose tissue of 5 healthy Holstein cows at 17 (±4 standard deviations) d postpartum. Cells were stimulated with increasing concentrations of ATRA (0.2, 2, and 20 nM) at the preconfluent (2 d) and postconfluent (8 d) preadipocyte stage or at the mature adipocyte stage (2 d). All concentrations of ATRA inhibited preconfluent preadipocyte proliferation with decreased proportion of S-phase cells and reduced protein abundance of cyclins (CCND1, CCND2, CCND3, CCNE1) and cyclin-dependent kinases (CDK2, CDK4, CDK6). Compared with vehicle, ATRA treatment induced apoptosis in preconfluent preadipocytes. Additionally, ATRA (0.2, 2, and 20 nM) supplementation also inhibited differentiation of postconfluent preadipocytes through downregulation of protein abundance of PPARγ and C/EBPα. After induction of differentiation, basal lipolysis in mature adipocytes increased upon treatment with all concentrations of ATRA. However, data on phosphorylated hormone-sensitive lipase or PLIN1 indicated that ATRA had no effect on epinephrine-stimulated lipolysis in mature adipocytes. Overall, these results demonstrate that ATRA might inhibit lipid accumulation by suppressing preadipocyte proliferation and differentiation, subsequently leading to apoptosis in postconfluent preadipocytes and promoting basal lipolysis in mature adipocytes. Overall, these in vitro responses provide some insights into the potential for nutritional management to modulate adipose tissue lipolysis, particularly in overconditioned cows during the dry period, which are more susceptible to suffer metabolic disorders due to excessive fat mobilization postpartum.
Sigma-Aldrich
Free Glycerol Determination Kit, 1 kit sufficient for 1000 reactions
Sigma-Aldrich
Anti-PKA Antibody, NT, Upstate®, from rabbit