MilliporeSigma
  • Radical scavenging and cytochrome P450 3A4 inhibitory activity of bergaptol and geranylcoumarin from grapefruit.
Radical scavenging and cytochrome P450 3A4 inhibitory activity of bergaptol and geranylcoumarin from grapefruit.
Bioorganic & medicinal chemistry (2007-04-03)
Basavaraj Girennavar, G K Jayaprakasha, Y Jadegoud, G A Nagana Gowda, Bhimanagouda S Patil
ABSTRACT
Grapefruit juice has been shown to increase the oral bioavailability of several clinically important drugs by inhibiting first pass metabolism. Several compounds in grapefruit juice have shown different biological activities. Unique among them are furocoumarins with potent inhibitory activity against cytochrome P450 enzymes. In the present study, two bioactive compounds were isolated from grapefruit juice and grapefruit peel oil. The purity of the isolated compounds has been analyzed by HPLC. Structures of the compounds were elucidated by extensive NMR and mass spectral studies and identified as bergaptol and geranylcoumarin. The isolated compounds were tested for their radical scavenging activity using 2,2'-azobis (3-ethylbenz-thiazoline-6-sulfonic acid) (ABTS) and 2,2-diphenyl-1-picrylhydrazil (DPPH) methods at different concentrations. Bergaptol showed very good radical scavenging activity at all the tested concentrations. Furthermore, these compounds were evaluated for their inhibitory activity against CYP3A4 enzyme. Bergaptol and geranylcoumarin were found to be potent inhibitors of debenzylation activity of CYP3A4 enzyme with an IC(50) value of 24.92 and 42.93 microM, respectively.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%