MilliporeSigma
  • Time-gated flow cytometry: an ultra-high selectivity method to recover ultra-rare-event mu-targets in high-background biosamples.
Time-gated flow cytometry: an ultra-high selectivity method to recover ultra-rare-event mu-targets in high-background biosamples.
Journal of biomedical optics (2009-05-02)
Dayong Jin, James A Piper, Robert C Leif, Sean Yang, Belinda C Ferrari, Jingli Yuan, Guilan Wang, Lidia M Vallarino, John W Williams
ABSTRACT
A fundamental problem for rare-event cell analysis is auto-fluorescence from nontarget particles and cells. Time-gated flow cytometry is based on the temporal-domain discrimination of long-lifetime (>1 micros) luminescence-stained cells and can render invisible all nontarget cell and particles. We aim to further evaluate the technique, focusing on detection of ultra-rare-event 5-microm calibration beads in environmental water dirt samples. Europium-labeled 5-microm calibration beads with improved luminescence homogeneity and reduced aggregation were evaluated using the prototype UV LED excited time-gated luminescence (TGL) flow cytometer (FCM). A BD FACSAria flow cytometer was used to sort accurately a very low number of beads (<100 events), which were then spiked into concentrated samples of environmental water. The use of europium-labeled beads permitted the demonstration of specific detection rates of 100%+/-30% and 91%+/-3% with 10 and 100 target beads, respectively, that were mixed with over one million nontarget autofluorescent background particles. Under the same conditions, a conventional FCM was unable to recover rare-event fluorescein isothiocyanate (FITC) calibration beads. Preliminary results on Giardia detection are also reported. We have demonstrated the scientific value of lanthanide-complex biolabels in flow cytometry. This approach may augment the current method that uses multifluorescence-channel flow cytometry gating.
Sigma-Aldrich
Goat Anti-Mouse light chain Antibody, biotin conjugate, Chemicon®, from goat