MilliporeSigma
  • Influence of Ligand and Nuclearity on the Cytotoxicity of Cyclometallated C^N^C Platinum(II) Complexes.
Influence of Ligand and Nuclearity on the Cytotoxicity of Cyclometallated C^N^C Platinum(II) Complexes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-06-11)
Angélique Kergreis, Rianne M Lord, Sarah J Pike
ABSTRACT
A series of cyclometallated mono- and di-nuclear platinum(II) complexes and the parent organic ligand, 2,6-diphenylpyridine 1 (HC^N^CH), have been synthesized and characterized. This library of compounds includes [(C^N^C)PtII (L)] (L=dimethylsulfoxide (DMSO) 2 and triphenylphosphine (PPh3 ) 3) and [((C^N^C)PtII )2 (L')] (where L'=N-heterocycles (pyrazine (pyr) 4, 4,4'-bipyridine (4,4'-bipy) 5 or diphosphine (1,4-bis(diphenylphosphino)butane (dppb) 6). Their cytotoxicity was assessed against four cancerous cell lines and one normal cell line, with results highlighting significantly increased antiproliferative activity for the dinuclear complexes (4-6), when compared to the mononucleated species (2 and 3). Complex 6 is the most promising candidate, displaying very high selectivity towards cancerous cells, with selectivity index (SI) values >29.5 (A2780) and >11.2 (A2780cisR), and outperforming cisplatin by >4-fold and >18-fold, respectively.
Sigma-Aldrich
2,6-Dibromopyridine, 98%