MilliporeSigma
  • Nephrotic syndrome is associated with increased plasma K+ concentration, intestinal K+ losses, and attenuated urinary K+ excretion: a study in rats and humans.
Nephrotic syndrome is associated with increased plasma K+ concentration, intestinal K+ losses, and attenuated urinary K+ excretion: a study in rats and humans.
American journal of physiology. Renal physiology (2019-10-01)
Rikke Ydegaard, Per Svenningsen, Claus Bistrup, Rene Frydensbjerg Andersen, Jane Stubbe, Kristian Bergholt Buhl, Niels Marcussen, Gitte Rye Hinrichs, Hiba Iraqi, Reza Zamani, Henrik Dimke, Boye L Jensen
ABSTRACT
The present study tested the hypotheses that nephrotic syndrome (NS) leads to renal K+ loss because of augmented epithelial Na+ channel (ENaC) activity followed by downregulation of renal K+ secretory pathways by suppressed aldosterone. The hypotheses were addressed by determining K+ balance and kidney abundance of K+ and Na+ transporter proteins in puromycin aminonucleoside (PAN)-induced rat nephrosis. The effects of amiloride and angiotensin II type 1 receptor and mineralocorticoid receptor (MR) antagonists were tested. Glucocorticoid-dependent MR activation was tested by suppression of endogenous glucocorticoid with dexamethasone. Urine and plasma samples were obtained from pediatric patients with NS in acute and remission phases. PAN-induced nephrotic rats had ENaC-dependent Na+ retention and displayed lower renal K+ excretion but elevated intestinal K+ secretion that resulted in less cumulated K+ in NS. Aldosterone was suppressed at day 8. The NS-associated changes in intestinal, but not renal, K+ handling responded to suppression of corticosterone, whereas angiotensin II type 1 receptor and MR blockers and amiloride had no effect on urine K+ excretion during NS. In PAN-induced nephrosis, kidney protein abundance of the renal outer medullary K+ channel and γ-ENaC were unchanged, whereas the Na+-Cl- cotransporter was suppressed and Na+-K+-ATPase increased. Pediatric patients with acute NS displayed suppressed urine Na+-to-K+ ratios compared with remission and elevated plasma K+ concentration, whereas fractional K+ excretion did not differ. Acute NS is associated with less cumulated K+ in a rat model, whereas patients with acute NS have elevated plasma K+ and normal renal fractional K+ excretion. In NS rats, K+ balance is not coupled to ENaC activity but results from opposite changes in renal and fecal K+ excretion with a contribution from corticosteroid MR-driven colonic secretion.
Sigma-Aldrich
Anti-SLC12A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SLC12A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Na+/K+ ATPase α-1 Antibody, Upstate®, from rabbit