MilliporeSigma
  • Platelet-activating factor (PAF) receptor binding antagonists from Alpinia officinarum.
Platelet-activating factor (PAF) receptor binding antagonists from Alpinia officinarum.
Bioorganic & medicinal chemistry letters (2007-10-30)
Gao-Jun Fan, Young-Hwa Kang, Yong Nam Han, Byung Hoon Han
ABSTRACT
The bioassay-guided purification of ether extracts of Alpinia officinarum led to the isolation of two new compounds 6-hydroxy-1,7-diphenyl-4-en-3-heptanone (1) and 6-(2-hydroxy-phenyl)-4-methoxy-2-pyrone (4) as well as three known compounds 1,7-diphenyl-4-en-3-heptanone (2), 1,7-diphenyl-5-methoxy-3-heptanone (3), and apigenin (5). Their structures were established on the basis of spectral methods. All three diarylheptanoids 1, 2, and 3 exhibited potent PAF receptor binding inhibitory activities with an IC(50) of 1.3, 5.0, and 1.6 microM, respectively. These studies have identified diarylheptanoids as a novel class of potent PAF antagonists.
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)