MilliporeSigma
  • Carbonic anhydrase activators: L-Adrenaline plugs the active site entrance of isozyme II, activating better isoforms I, IV, VA, VII, and XIV.
Carbonic anhydrase activators: L-Adrenaline plugs the active site entrance of isozyme II, activating better isoforms I, IV, VA, VII, and XIV.
Bioorganic & medicinal chemistry letters (2006-11-28)
Claudia Temperini, Alessio Innocenti, Andrea Scozzafava, Antonio Mastrolorenzo, Claudiu T Supuran
ABSTRACT
The activation of the metalloenzyme carbonic anhydrase (CA, EC 4.2.1.1) with L-adrenaline and histamine has been investigated by kinetic and X-ray crystallographic studies. L-Adrenaline behaves as a potent activator of isozyme CA I (activation constant of 90 nM), being a much weaker activator of isozyme CA II (activation constant of 96 microM). Isoforms CA IV, VA, VII, and XIV were activated by L-adrenaline with K(A)s in the range of 36-63 microM. The X-ray crystal structure of the CA II-L-adrenaline adduct revealed that the activator plugs the entrance of the active site cavity, obstructing it almost completely.
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
(−)-Epinephrine