MilliporeSigma
  • Selinexor Sensitizes TRAIL-R2-Positive TNBC Cells to the Activity of TRAIL-R2xCD3 Bispecific Antibody.
Selinexor Sensitizes TRAIL-R2-Positive TNBC Cells to the Activity of TRAIL-R2xCD3 Bispecific Antibody.
Cells (2020-10-08)
Silvia Martini, Mariangela Figini, Aurora Croce, Barbara Frigerio, Marzia Pennati, Alessandro Massimo Gianni, Cinzia De Marco, Maria Grazia Daidone, Christian Argueta, Yosef Landesman, Nadia Zaffaroni, Alessandro Satta
ABSTRACT
Triple-negative breast cancer (TNBC) is an aggressive disease with poor prognosis and limited therapeutic options. Recent advances in the immunotherapy field have enabled the development of new treatment strategies, among which the use of bispecific antibodies (BsAbs), able to redirect T cells against tumors, has shown promising results. In particular, a BsAb that uses TNF-related apoptosis-inducing ligand receptor 2 (TRAIL-R2) as a target was constructed and demonstrated good results in redirecting CD3+ T cells to kill TRAIL-R2-expressing TNBC cells. In the present study, we investigated whether treatment with selinexor, a selective inhibitor of nuclear export (SINE) targeting exportin-1/chromosome maintenance protein 1 (XPO1/CRM1), could potentiate the antitumor activity of this BsAb. In combination experiments, we found that selinexor-exposed TNBC cells exhibited greater growth inhibition when treated with the TRAIL-R2xCD3 BsAb than that expected by simple additivity. Similarly, the apoptosis rate in selinexor/TRAIL-R2xCD3 BsAb-treated TNBC cells was significantly higher than that observed after exposure to either single agent. Together, our results suggest that the combination of selinexor and TRAIL-R2xCD3 BsAb can be a viable anticancer strategy and indicate this treatment as a promising therapeutic option for TNBC patients.
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFRSF10B
Sigma-Aldrich
Anti-TNFRSF10B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution