MilliporeSigma
  • Transducin β-like protein 1 controls multiple oncogenic networks in diffuse large B-cell lymphoma.
Transducin β-like protein 1 controls multiple oncogenic networks in diffuse large B-cell lymphoma.
Haematologica (2020-10-16)
Youssef Youssef, Vrajesh Karkhanis, Wing Keung Chan, Frankie Jeney, Alessandro Canella, Xiaoli Zhang, Shelby Sloan, Alexander Prouty, JoBeth Helmig-Mason, Liudmyla Tsyba, Walter Hanel, Xuguang Zheng, Pu Zhang, Ji-Hyun Chung, David M Lucas, Zachary Kauffman, Karilyn Larkin, Anne M Strohecker, Hatice G Ozer, Rosa Lapalombella, Hui Zhou, Zijun Y Xu-Monette, Ken H Young, Ruolan Han, Elmar Nurmemmedov, Gerard Nuovo, Kami Maddocks, John C Byrd, Robert A Baiocchi, Lapo Alinari
ABSTRACT
Diffuse large B-cell lymphoma (DLBCL) is the most common Non-Hodgkin's lymphoma and is characterized by a remarkable heterogeneity with diverse variants that can be identified histologically and molecularly. Large-scale gene expression profiling studies have identified the germinal center B-cell (GCB-) and activated B-cell (ABC-) subtypes. Standard chemo-immunotherapy remains standard front line therapy, curing approximately two thirds of patients. Patients with refractory disease or those who relapse after salvage treatment have an overall poor prognosis highlighting the need for novel therapeutic strategies. Transducin β-like protein 1 (TBL1) is an exchange adaptor protein encoded by the TBL1X gene and known to function as a master regulator of the Wnt signalling pathway by binding to β-CATENIN and promoting its downstream transcriptional program. Here, we show that, unlike normal B-cells, DLBCL cells express abundant levels of TBL1 and its overexpression correlates with poor clinical outcome regardless of DLBCL molecular subtype. Genetic deletion of TBL1 and pharmacological approach using tegavivint, a first-in-class small molecule targeting TBL1 (Iterion Therapeutics), promotes DLBCL cell death in vitro and in vivo. Through an integrated genomic, biochemical, and pharmacologic analyses, we characterized a novel, β-CATENIN independent, post-transcriptional oncogenic function of TBL1 in DLBCL where TBL1 modulates the stability of key oncogenic proteins such as PLK1, MYC, and the autophagy regulatory protein BECLIN-1 through its interaction with a SKP1-CUL1-F-box (SCF) protein supercomplex. Collectively, our data provide the rationale for targeting TBL1 as a novel therapeutic strategy in DLBCL.
Sigma-Aldrich
Anti-TBL1X antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CUL1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CAND1, (N-terminal) antibody produced in mouse, clone 4D10, purified immunoglobulin, buffered aqueous solution