MilliporeSigma
  • Matrix-dependent a/x pair and overdegraded w/y/z ions generated by radical-directed dissociation of peptide radical cations [M]+ in matrix-assisted laser desorption/ionization in-source decay.
Matrix-dependent a/x pair and overdegraded w/y/z ions generated by radical-directed dissociation of peptide radical cations [M]+ in matrix-assisted laser desorption/ionization in-source decay.
Journal of mass spectrometry : JMS (2020-11-03)
Kei Miyazawa, Mitsuo Takayama
ABSTRACT
Fragmentation of peptide radical cations [M]. + has been examined using matrix-assisted laser desorption/ionization (MALDI) in-source decay (ISD) with hydrogen-abstracting nitro-substituted matrices. The ISD spectra of peptides containing an arginine (Arg) residue at carboxyl (C)-termini showed preferential [w]+ ions when 4-nitro-1-naphthol (4,1-NNL) matrix was used, whereas the use of 3,5-dinitrosalicylic acid (3,5-DNSA) resulted in preferential [x]+ ions. Minor or some [d]+ , [x]+ , [y]+ , and [z]+ ions were also observed. For peptides containing Arg residue at amino (N)-termini, the ISD spectra showed preferential [a]+ ions independent of matrix used. The observed [a]+ , [w]+ , [x]+ , [y]+ , and [z]+ ions can be rationally explained by radical-directed dissociation (RDD) of the peptide radical cations [M]. + , although [d]+ ions may be formed via Norrish Type I cleavage and/or by RDD of [M]. + ions. The formation of overdegraded [d]+ , [w]+ , [y]+ , and [z]+ ions is discussed from the standpoint of the internal energy of radical cations [M]. + and radical fragment ions [a + H]. + and [x + H]. + deposited via collisional interactions with excited matrix molecules in the MALDI plume. The radical site of the peptide cations [M]. + was presumed to be backbone amide nitrogen, from MALDI-ISD data with three different deuterated amino acids.