MilliporeSigma
  • Personalized Identification of Optimal HIPEC Perfusion Protocol in Patient-Derived Tumor Organoid Platform.
Personalized Identification of Optimal HIPEC Perfusion Protocol in Patient-Derived Tumor Organoid Platform.
Annals of surgical oncology (2020-07-08)
Steven D Forsythe, Shyama Sasikumar, Omeed Moaven, Hemamylammal Sivakumar, Perry Shen, Edward A Levine, Shay Soker, Aleksander Skardal, Konstantinos I Votanopoulos
ABSTRACT
Chemotherapy dosing duration and perfusion temperature vary significantly in HIPEC protocols. This study investigates patient-derived tumor organoids as a platform to identify the most efficacious perfusion protocol in a personalized approach. Peritoneal tumor tissue from 15 appendiceal and 8 colon cancer patients who underwent CRS/HIPEC were used for personalized organoid development. Organoids were perfused in parallel at 37 and 42 °C with low- and high-dose oxaliplatin (200 mg/m2 over 2 h vs. 460 mg/m2 over 30 min) and MMC (40 mg/3L over 2 h). Viability assays were performed and pooled for statistical analysis. An adequate organoid number was generated for 75% (6/8) of colon and 73% (11/15) of appendiceal patients. All 42 °C treatments displayed lower viability than 37 °C treatments. On pooled analysis, MMC and 200 mg/m2 oxaliplatin displayed no treatment difference for either appendiceal or colon organoids (19% vs. 25%, p = 0.22 and 27% vs. 31%, p = 0.55, respectively), whereas heated MMC was superior to 460 mg/m2 oxaliplatin in both primaries (19% vs. 54%, p < 0.001 and 27% vs. 53%, p = 0.002, respectively). In both appendiceal and colon tumor organoids, heated 200 mg/m2 oxaliplatin displayed increased cytotoxicity as compared with 460 mg/m2 oxaliplatin (25% vs. 54%, p < 0.001 and 31% vs. 53%, p = 0.008, respectively). Organoids treated with MMC or 200 mg/m2 heated oxaliplatin for 2 h displayed increased susceptibility in comparison with 30-min 460 mg/m2 oxaliplatin. Optimal perfusion protocol varies among patients, and organoid technology may offer a platform for tailoring HIPEC conditions to the individual patient level.
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Gentamicin solution, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
A 83-01, ≥98% (HPLC)
Millipore
Millipore® Steriflip® Vacuum Tube Top Filter, pore size 100.0 μm, Nylon membrane (net), funnel capacity 50 mL, pack of 25 ea
Sigma-Aldrich
Folinic acid calcium salt hydrate, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor