MilliporeSigma
Synthesis and biological evaluation of 12 allenic aromatic ethers.
Bioorganic & medicinal chemistry letters (2007-03-28)
San-Yong Wang, Wei-Wei Mao, Zhi-Gang She, Chun-Rong Li, Ding-Qiao Yang, Yong-Cheng Lin, Li-Wu Fu
ABSTRACT
Twelve allenic aromatic ethers, some of them are natural products isolated from the mangrove fungus Xylaria sp. 2508 in the South China Sea, were synthesized. Their antitumor activities against KB and KBv200 cells were determined. All these compounds demonstrated cytotoxic potential, ranging from weak to strong activity. The analysis of structure-activity relationships suggested that the introduction of allenic moiety could generate or enhance cytotoxicity of these phenol compounds.
Product Description
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzyl alcohol, ≥98%, FG
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.