MilliporeSigma
  • Recruitment of activated G protein-coupled receptors to pre-existing clathrin-coated pits in living cells.
Recruitment of activated G protein-coupled receptors to pre-existing clathrin-coated pits in living cells.
The Journal of biological chemistry (2001-10-17)
Mark G H Scott, Alexandre Benmerah, Olivier Muntaner, Stefano Marullo
ABSTRACT
The process of clathrin-mediated endocytosis tightly regulates signaling of the superfamily of seven-transmembrane G protein-coupled receptors (GPCRs). A fundamental question in the cell biology of membrane receptor endocytosis is whether activated receptors can initiate the formation of clathrin-coated pits (CPs) or whether they are simply mobilized to pre-existing CPs. Here, using various approaches, including a dynamic assay to monitor the distribution of CPs and GPCR-beta-arrestin complexes in live HeLa cells, we demonstrate for the first time that activated GPCRs do not initiate the de novo formation of CPs but instead are targeted to pre-existing CPs.
Sigma-Aldrich
Monoclonal Anti-α-Adaptin antibody produced in mouse, clone 100/2, ascites fluid