MilliporeSigma
  • Synthesis of new sugar derivatives from Stachys sieboldi Miq and antibacterial evaluation against Mycobacterium tuberculosis, Mycobacterium avium, and Staphylococcus aureus.
Synthesis of new sugar derivatives from Stachys sieboldi Miq and antibacterial evaluation against Mycobacterium tuberculosis, Mycobacterium avium, and Staphylococcus aureus.
Bioorganic & medicinal chemistry letters (2007-03-03)
Taku Chiba, Takemasa Takii, Kenji Nishimura, Yoshifumi Yamamoto, Hiroko Morikawa, Chiyoji Abe, Kikuo Onozaki
ABSTRACT
A series of sugar derivatives (7-14) were synthesized from stachyose, a sugar compound of Stachys sieboldi Miq, and evaluated for antibacterial activity against Mycobacterium tuberculosis, Mycobacterium avium, and Staphylococcus aureus, and their structure-activity relationships were studied. The results showed that the compound OCT359 (allyl O-(2,3,4,6-tetra-O-acetyl-alpha-D-galactopyranosyl)-(1-->6)-O-(2,3,4-tri-O-acetyl-alpha-D-galactopyranosyl)-(1-->6)-O-2,3,4-tri-O-acetyl-beta-D-glucopyranoside) (12) exhibited in vitro antibacterial activity. The allyl group at C-1 and the acetoxy groups of the manninotrioside were requisite for the antibacterial activity.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Allyl isothiocyanate, 95%
Sigma-Aldrich
Allyl isothiocyanate, ≥95%, FCC
Supelco
Allyl isothiocyanate, PESTANAL®, analytical standard
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Sigma-Aldrich
Allyl alcohol, ≥99%