MilliporeSigma
  • Beta3-adrenoceptor is the predominant beta-adrenoceptor subtype in human myometrium and its expression is up-regulated in pregnancy.
Beta3-adrenoceptor is the predominant beta-adrenoceptor subtype in human myometrium and its expression is up-regulated in pregnancy.
The Journal of clinical endocrinology and metabolism (2004-12-09)
C Rouget, M Bardou, M Breuiller-Fouché, C Loustalot, H Qi, E Naline, T Croci, D Cabrol, C Advenier, M J Leroy
ABSTRACT
To assess whether pregnancy might influence the functionality and expression of human myometrial beta(2)- and beta(3)-adrenoceptors (beta(2)- and beta(3)-AR), we performed functional, binding, Western blot, and molecular biology experiments in human nonpregnant and near-term pregnant myometrium. Inhibition of spontaneous contractions induced by a beta(3)-AR agonist, SR 59119A, was significantly greater in pregnant, compared with nonpregnant, myometrial strips (E'(max) = 61 +/- 5% vs. 44 +/- 5% for pregnant and nonpregnant myometrium, respectively), whereas salbutamol, a beta(2)-AR agonist, was significantly less efficient in pregnant, compared with nonpregnant, myometrium (E(max) = 29 +/- 4 vs. 54 +/- 8%). Although two populations of binding sites corresponding to beta(2)- and beta(3)-AR were identified in both nonpregnant and pregnant myometrium, we found a clear predominance of the beta(3)-AR subtype. Moreover, beta(3)-AR binding sites were up-regulated 2-fold in myometrium at the end of pregnancy. Both beta(2)- and beta(3)-AR mRNA were expressed in human nonpregnant and pregnant myometrium. Contrary to beta(2)-AR, the expression of the beta(3)-AR transcripts and immunoreactive proteins was increased in pregnant, compared with nonpregnant, myometrium. Such compelling data suggest a predominant role for beta(3)-AR in the regulation of human myometrium contractility, especially at the end of pregnancy, which might have important consequences for the clinical management of preterm labor.
Sigma-Aldrich
Anti-Adrenergic Receptor Antibody, β 3, Chemicon®, from rabbit