Merck
Food web-specific biomagnification of persistent organic pollutants.
Science (New York, N.Y.) (2007-07-14)
Barry C Kelly, Michael G Ikonomou, Joel D Blair, Anne E Morin, Frank A P C Gobas
ABSTRACT
Substances that accumulate to hazardous levels in living organisms pose environmental and human-health risks, which governments seek to reduce or eliminate. Regulatory authorities identify bioaccumulative substances as hydrophobic, fat-soluble chemicals having high octanol-water partition coefficients (K(OW))(>/=100,000). Here we show that poorly metabolizable, moderately hydrophobic substances with a K(OW) between 100 and 100,000, which do not biomagnify (that is, increase in chemical concentration in organisms with increasing trophic level) in aquatic food webs, can biomagnify to a high degree in food webs containing air-breathing animals (including humans) because of their high octanol-air partition coefficient (K(OA)) and corresponding low rate of respiratory elimination to air. These low K(OW)-high K(OA) chemicals, representing a third of organic chemicals in commercial use, constitute an unidentified class of potentially bioaccumulative substances that require regulatory assessment to prevent possible ecosystem and human-health consequences.
Product Description
2,2′,4,4′,5,5′-Hexachlorobiphenyl (IUPAC No. 153), BCR®, certified reference material
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Mirex, PESTANAL®, analytical standard
Supelco
PCB No 153, analytical standard
Supelco
Styrene, analytical standard
Supelco
β-HCH, PESTANAL®, analytical standard
Supelco
BDE No 99 solution, 50 μg/mL in isooctane, analytical standard
2,2′,3,4,4′,5,5′-Heptachlorobiphenyl (IUPAC No. 180), BCR®, certified reference material
Supelco
Tetradifon, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachlorobenzene, 96%
Sigma-Aldrich
1,2,4,5-Tetrachlorobenzene, 98%
Sigma-Aldrich
Hexachloro-1,3-butadiene, 96%
Supelco
Pentachlorobenzene, PESTANAL®, analytical standard
Supelco
PCB No 180, analytical standard
Supelco
Hexachlorobenzene, PESTANAL®, analytical standard