MilliporeSigma
  • Structure-activity relationship study of prion inhibition by 2-aminopyridine-3,5-dicarbonitrile-based compounds: parallel synthesis, bioactivity, and in vitro pharmacokinetics.
Structure-activity relationship study of prion inhibition by 2-aminopyridine-3,5-dicarbonitrile-based compounds: parallel synthesis, bioactivity, and in vitro pharmacokinetics.
Journal of medicinal chemistry (2007-01-05)
Barnaby C H May, Julie A Zorn, Juanita Witkop, John Sherrill, Andrew C Wallace, Giuseppe Legname, Stanley B Prusiner, Fred E Cohen
ABSTRACT
2-Aminopyridine-3,5-dicarbonitrile compounds were previously identified as mimetics of dominant-negative prion protein mutants and inhibit prion replication in cultured cells. Here, we report findings from a comprehensive structure-activity relationship study of the 6-aminopyridine-3,5-dicarbonitrile scaffold. We identify compounds with significantly improved bioactivity (approximately 40-fold) against replication of the infectious prion isoform (PrPSc) and suitable pharmacokinetic profiles to warrant evaluation in animal models of prion disease.
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Furosemide
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, ≥98%