MilliporeSigma
  • Anti-complement C5 therapy with eculizumab in three cases of critical COVID-19.
Anti-complement C5 therapy with eculizumab in three cases of critical COVID-19.
Clinical immunology (Orlando, Fla.) (2020-08-11)
Jeffrey Laurence, J Justin Mulvey, Madhav Seshadri, Alexandra Racanelli, Joanna Harp, Edward J Schenck, Dana Zappetti, Evelyn M Horn, Cynthia M Magro
ABSTRACT
Respiratory failure and acute kidney injury (AKI) are associated with high mortality in SARS-CoV-2-associated Coronavirus disease 2019 (COVID-19). These manifestations are linked to a hypercoaguable, pro-inflammatory state with persistent, systemic complement activation. Three critical COVID-19 patients recalcitrant to multiple interventions had skin biopsies documenting deposition of the terminal complement component C5b-9, the lectin complement pathway enzyme MASP2, and C4d in microvascular endothelium. Administration of anti-C5 monoclonal antibody eculizumab led to a marked decline in D-dimers and neutrophil counts in all three cases, and normalization of liver functions and creatinine in two. One patient with severe heart failure and AKI had a complete remission. The other two individuals had partial remissions, one with resolution of his AKI but ultimately succumbing to respiratory failure, and another with a significant decline in FiO2 requirements, but persistent renal failure. In conclusion, anti-complement therapy may be beneficial in at least some patients with critical COVID-19.
Sigma-Aldrich
Anti-MASP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution