MilliporeSigma
  • Impact of training state on fasting-induced regulation of adipose tissue metabolism in humans.
Impact of training state on fasting-induced regulation of adipose tissue metabolism in humans.
Journal of applied physiology (Bethesda, Md. : 1985) (2017-12-02)
Lærke Bertholdt, Anders Gudiksen, Tomasz Stankiewicz, Ida Villesen, Jonas Tybirk, Gerrit van Hall, Jens Bangsbo, Peter Plomgaard, Henriette Pilegaard
ABSTRACT
Recruitment of fatty acids from adipose tissue is increased during fasting. However, the molecular mechanisms behind fasting-induced metabolic regulation in human adipose tissue and the potential impact of training state in this are unknown. Therefore the aim of the present study was to investigate 1) fasting-induced regulation of lipolysis and glyceroneogenesis in human adipose tissue as well as 2) the impact of training state on basal oxidative capacity and fasting-induced metabolic regulation in human adipose tissue. Untrained [maximal oxygen uptake (V̇o2max) < 45 ml·min-1·kg-1] and trained subjects (V̇o2max > 55 ml·min-1·kg-1) fasted for 36 h, and abdominal subcutaneous adipose tissue biopsies were obtained 2, 12, 24, and 36 h after a standardized meal. Adipose tissue oxidative phosphorylation complexes, phosphoenolpyruvate carboxykinase, and pyruvate dehydrogenase (PDH)-E1α protein as well as PDH kinase (PDK) 2, PDK4, and PDH phosphatase 2 mRNA content were higher in trained subjects than in untrained subjects. In addition, trained subjects had higher adipose tissue hormone-sensitive lipase Ser660 phosphorylation and adipose triglyceride lipase protein content as well as higher plasma free fatty acid concentration than untrained subjects during fasting. Moreover, adipose tissue PDH phosphorylation increased with fasting only in trained subjects. Taken together, trained subjects seem to possess higher basal adipose tissue oxidative capacity as well as higher capacity for regulation of lipolysis and for providing substrate for glyceroneogenesis in adipose tissue during fasting than untrained subjects. NEW & NOTEWORTHY This study shows for the first time higher protein content of lipolytic enzymes and higher oxidative phosphorylation protein in adipose tissue from trained subjects than from untrained subjects during fasting. Furthermore, trained subjects had higher capacity for adipose tissue glyceroneogenesis than untrained subjects.
Sigma-Aldrich
PhosphoDetect Anti-PDH-E1α (pSer²³²) Rabbit pAb, liquid, Calbiochem®
Pricing and availability is not currently available.