MilliporeSigma
  • Synthesis and structure-activity relationship studies of 3,6-diazabicyclo[3.2.0]heptanes as novel alpha4beta2 nicotinic acetylcholine receptor selective agonists.
Synthesis and structure-activity relationship studies of 3,6-diazabicyclo[3.2.0]heptanes as novel alpha4beta2 nicotinic acetylcholine receptor selective agonists.
Journal of medicinal chemistry (2007-10-13)
Jianguo Ji, Michael R Schrimpf, Kevin B Sippy, William H Bunnelle, Tao Li, David J Anderson, Connie Faltynek, Carol S Surowy, Tino Dyhring, Philip K Ahring, Michael D Meyer
ABSTRACT
A series of novel, potent neuronal nicotinic acetylcholine receptor (nAChR) ligands derived from 3,6-diazabicyclo[3.2.0]heptane have been synthesized and evaluated for binding affinity and agonist activity at the alpha4beta2 nAChR subtype. Structure-activity relationship studies of these novel nAChR ligands focused on substitution effects on the pyridine ring, as well as stereo- and regiochemical influences of the 3,6-diazabicyclo[3.2.0]heptane core. Small 5-substituents on the pyridine ring had a modest impact on the binding affinities and functional activities. 6-Bromo, 6-chloro, and 6-methyl substituents on the pyridine ring led to increased binding affinities and improved functional activities. Most of the 6-N-pyridinyl-substituted 3,6-diazabicyclo[3.2.0]heptanes are selective for the alpha4beta2 nAChR subtype. Compounds (1R,5S)-25, (1R,5S)-55, and (1R,5S)-56 were virtually inactive as agonists at the halpha3beta4 nAChR but retained potency and efficacy at the halpha4beta2 nAChR subtype. 3-N-Pyridinyl-substituted series demonstrated more complex SAR. (1R,5R)-39, (1R,5R)-41, and (1R,5R)-42 were found to be much more potent at the halpha3beta4 nAChR subtype, whereas (1R,5R)-38 and (1R,5R)-40 were very selective at the halpha4beta2 nAChR subtype. The SAR studies of these novel ligands led to the discovery of several compounds with interesting in vitro pharmacological profiles.
Product Description
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid