MilliporeSigma
  • Design, synthesis, and evaluation of inhibitors for severe acute respiratory syndrome 3C-like protease based on phthalhydrazide ketones or heteroaromatic esters.
Design, synthesis, and evaluation of inhibitors for severe acute respiratory syndrome 3C-like protease based on phthalhydrazide ketones or heteroaromatic esters.
Journal of medicinal chemistry (2007-03-27)
Jianmin Zhang, Hanna I Pettersson, Carly Huitema, Chunying Niu, Jiang Yin, Michael N G James, Lindsay D Eltis, John C Vederas
ABSTRACT
The 3C-like protease (3CLpro), which controls the severe acute respiratory syndrome (SARS) coronavirus replication, has been identified as a potential target for drug design in the treatment of SARS. A series of tetrapeptide phthalhydrazide ketones, pyridinyl esters, and their analogs have been designed, synthesized, and evaluated as potential SARS 3CLpro inhibitors. Some pyridinyl esters are identified as very potent inhibitors, with IC50 values in the nanomolar range (50-65 nM). Electrospray mass spectrometry indicates a mechanism involving acylation of the active site cysteine thiol for this class of inhibitors.
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Furfural, 99%