MilliporeSigma
  • Interaction of Trichomonas vaginalis and Tritrichomonas foetus with keratin: an important role in parasite infection.
Interaction of Trichomonas vaginalis and Tritrichomonas foetus with keratin: an important role in parasite infection.
Memorias do Instituto Oswaldo Cruz (2011-10-21)
Ricardo Chaves Vilela, Marlene Benchimol
ABSTRACT
Trichomonas vaginalis and Tritrichomonas foetus are human and bovine parasites, respectively, that provoke the sexually transmitted disease trichomoniasis. These extracellular parasites adhere to the host epithelial cell surface. Although mucinases and proteases have been described as important proteins for parasite adhesion to epithelial cells, no studies have examined the role of the keratin molecules that cornify the vaginal epithelium. Here, we investigated the interaction of T. vaginalis and T. foetus with human keratin in vitro; additionally, adherence assays were performed in cattle with T. foetus to elucidate whether trichomonads were able to interact with keratin in vivo. We demonstrated that both T. vaginalisand T. foetusinteracted directly with keratin. Additionally, the trichomonads ingested and digested keratin, shedding new light on the Trichomonas infection process.
Sigma-Aldrich
Anti-Cytokeratin, pan antibody, Mouse monoclonal, clone PCK-26, purified from hybridoma cell culture