MilliporeSigma
  • MiR-22-3p inhibits fibrotic cataract through inactivation of HDAC6 and increase of α-tubulin acetylation.
MiR-22-3p inhibits fibrotic cataract through inactivation of HDAC6 and increase of α-tubulin acetylation.
Cell proliferation (2020-09-29)
Xiaoran Wang, Liping Wang, Yan Sun, Baoxin Chen, Lang Xiong, Jieping Chen, Mi Huang, Jing Wu, Xuhua Tan, Yingfeng Zheng, Shan Huang, Yizhi Liu
ABSTRACT
Fibrotic cataract, including posterior capsule opacification (PCO) and anterior subcapsular cataract (ASC), renders millions of people visually impaired worldwide. However, the underlying mechanism remains poorly understood. Here, we report a miRNA-based regulatory pathway that controls pathological fibrosis of lens epithelium. Expression of miR-22-3p and histone deacetylase 6 (HDAC6) in normal and PCO patient samples were measured by qPCR. Human lens epithelial explants were treated with TGF-β2 in the presence or absence of miR-22-3p mimics or inhibitor. Cell proliferation was determined by MTS assay, and migration was tested by transwell assay. Expression of HDAC6 and EMT-related molecules were analysed by Western blot, qPCR and immunocytochemical experiments. We identify miR-22-3p as a downregulated miRNA targeting HDAC6 in LECs during lens fibrosis and TGF-β2 treatment. Mechanistically, gain- and loss-of-function experiments in human LECs and lens epithelial explants reveal that miR-22-3p prevents proliferation, migration and TGF-β2 induced EMT of LECs via targeting HDAC6 and thereby promoting α-tubulin acetylation. Moreover, pharmacological targeting of HDAC6 deacetylase with Tubacin prevents fibrotic opaque formation through increasing α-tubulin acetylation under TGF-β2 stimulated conditions in both human lens epithelial explants and the whole rat lenses. These findings suggest that miR-22-3p prevents lens fibrotic progression by targeting HDAC6 thereby promoting α-tubulin acetylation. The 'miR-22-HDAC6-α-tubulin (de)acetylation' signalling axis may be therapeutic targets for the treatment of fibrotic cataract.
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-HDAC6 Antibody, CT, Upstate®, from rabbit