MilliporeSigma
  • Adenovirus-mediated gene transfer of macrophage colony stimulating factor to the arterial wall in vivo.
Adenovirus-mediated gene transfer of macrophage colony stimulating factor to the arterial wall in vivo.
Arteriosclerosis, thrombosis, and vascular biology (1998-07-22)
G Mozes, T Mohacsi, P Gloviczki, S Menawat, I Kullo, D Spector, J Taylor, T B Crotty, T O'Brien
ABSTRACT
Macrophage colony stimulating factor (MCSF) is believed to play a key role in one of the earliest events in atherosclerosis, ie, monocyte to macrophage differentiation in the arterial intima. The aim of this study was to examine the biological effects of vascular wall expression of MCSF. A recombinant adenovirus vector encoding human MCSF (AdMCSF) was generated by standard techniques of homologous recombination in 293 cells. The rabbit carotid artery was transduced with AdMCSF. As negative controls, carotid arteries were transduced with either an adenoviral vector encoding beta-galactosidase, an adenoviral vector encoding apolipoprotein E, or diluent alone. Intima-media thickness ratio was calculated 5 and 21 days after transduction. The cell type present in intimal infiltrates was analyzed by immunohistochemistry. MCSF expression was demonstrated in the vessel wall of AdMCSF-transduced vessels by reverse transcription-polymerase chain reaction and immunofluorescence. In contrast to control vessels, adenovirus-mediated MCSF expression was associated with an intimal cellular infiltrate consisting of smooth muscle cells and small numbers of macrophages. Whereas the intima-media thickness ratio was greater in AdMCSF-transduced vessels at 5 days, this difference was no longer statistically significant at 21 days. These results suggest that MCSF may play a role in recruitment of monocytes and macrophages to the vessel wall and may contribute to smooth muscle cell proliferation and migration.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−TRITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution