MilliporeSigma
[Effect of irisin on hypoxic-ischemic brain damage in neonatal rats].
Zhongguo dang dai er ke za zhi = Chinese journal of contemporary pediatrics (2020-01-18)
Xuan-Pei Xu, Ling-Yi Huang, Feng-Yan Zhao, Jun-Jie Ying, Shi-Ping Li, Yan Yue, Wen-Xing Li, Yi Qu, De-Zhi Mu
ABSTRACT
To study the effect and mechanism of action of irisin on hypoxic-ischemic brain damage in neonatal rats. A total of 248 7-day-old Sprague-Dawley rats were randomly divided into a sham-operation group, a model group, and low- and high-dose irisin intervention groups (n=62 each). The rats in the model and irisin intervention groups were given hypoxic treatment after right common carotid artery ligation to establish a model of hypoxic-ischemic brain damage. Those in the sham-operation group were given the separation of the right common carotid artery without ligation or hypoxic treatment. The rats in the high- and low-dose irisin intervention groups were given intracerebroventricular injection of recombinant irisin polypeptide at a dose of 0.30 µg and 0.15 µg respectively. Those in the model and sham-operation groups were given the injection of an equal volume of PBS. The water maze test was used to compare neurological behaviors between groups. TTC staining, hematoxylin-eosin staining and TUNEL staining were used to observe histopathological changes of the brain. Western blot was used to measure the expression of the apoptosis-related molecules cleaved-caspase-3 (CC3), BCL-2 and BAX. Compared with the sham-operation group, the model group had a significant increase in latency time and a significant reduction in the number of platform crossings (P<0.05). Compared with the model group, the high-dose irisin intervention group had a significant reduction in latency time and a significant increase in the number of platform crossings (P<0.05). Compared with the sham-operation group, the model group had massive infarction in the right hemisphere, with significant increases in karyopyknosis and karyorrhexis. Compared with the model group, the high-dose irisin intervention group had a smaller infarct area of the right hemisphere, with reductions in karyopyknosis and karyorrhexis. The model group had a significantly higher apoptosis rate of cells in the right cerebral cortex and the hippocampus than the sham-operation group. The high-dose irisin intervention group had a significantly lower apoptosis rate than the model group (P<0.05). At 24 and 48 hours after modeling, the sham-operation group had a significantly lower level of CC3 than the model group (P<0.05). Compared with the model group, the high-dose irisin intervention group had a significantly lower level of CC3 and a significantly higher BCL-2/BAX ratio (P<0.05). The low-dose irisin intervention group had similar laboratory markers and histopathological changes of the brain to the model group. Irisin can alleviate hypoxic-ischemic brain damage in neonatal rats in a dose-dependent manner, possibly by reducing cell apoptosis in the cerebral cortex and the hippocampus.
Sigma-Aldrich
Anti-BCL-2 antibody produced in rabbit, affinity isolated antibody
Roche
In Situ Cell Death Detection Kit, POD, sufficient for ≤50 tests