MilliporeSigma
  • Experimental infection of inbred BALB/c and A/J mice with Massachusetts and Brazilian strains of infectious bronchitis virus (IBV).
Experimental infection of inbred BALB/c and A/J mice with Massachusetts and Brazilian strains of infectious bronchitis virus (IBV).
Archives of virology (2015-05-09)
Matheus C Martini, Jacy Gameiro, Tereza Cristina Cardoso, Leonardo C Caserta, Ana Carolina M Gualberto, Lara Borges Keid, Trícia M F de S Oliveira, Marcia M A B dos Santos, Clarice W Arns, Helena L Ferreira
ABSTRACT
The ability of avian coronaviruses to replicate in mice was investigated to investigate interspecies transmission. Two inbred mouse strains (BALB/c and A/J) with different genetic backgrounds were inoculated with the avian coronavirus strains Mass and BR-I and monitored for at least 10 days. Analysis of viral RNA, histopathological examinations, immunohistochemistry and serology were performed. After virus inoculation, neither clinical signs nor evident gross lesions were observed. Viral RNA, histopathological changes, and viral nucleoprotein were observed in the lung, trachea and sinus of all inoculated mice. Our study demonstrates the importance of elucidating the epidemiology of coronaviruses, including in rodents that are pests in poultry production.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution