MilliporeSigma
  • Erratum: Overexpression of a fusion defensin gene from radish and fenugreek improves resistance against leaf spot diseases caused by Cercospora arachidicola and Phaeoisariopsis personata in peanut.
Erratum: Overexpression of a fusion defensin gene from radish and fenugreek improves resistance against leaf spot diseases caused by Cercospora arachidicola and Phaeoisariopsis personata in peanut.
Turkish journal of biology = Turk biyoloji dergisi (2019-08-15)
Madhu Bala, Thankappan Radhakrishnan, Abhay Kumar, Gyan Prakash Mishra, Jentilal Ramjibhai Dobraia, Pulugurtha Bharadwaja Kirti
ABSTRACT
[This corrects the article DOI: 10.3906/biy-1412-46.].
Sigma-Aldrich
Palladium(II) acetylacetonate, ≥99.9% trace metals basis