MilliporeSigma
  • The RECAP Test Rapidly and Reliably Identifies Homologous Recombination-Deficient Ovarian Carcinomas.
The RECAP Test Rapidly and Reliably Identifies Homologous Recombination-Deficient Ovarian Carcinomas.
Cancers (2020-10-03)
Lise M van Wijk, Sylvia Vermeulen, Matty Meijers, Manuela F van Diest, Natalja T Ter Haar, Marthe M de Jonge, Nienke Solleveld-Westerink, Tom van Wezel, Dik C van Gent, Judith R Kroep, Tjalling Bosse, Katja N Gaarenstroom, Harry Vrieling, Maaike P G Vreeswijk
ABSTRACT
Recent studies have shown that the efficacy of PARP inhibitors in epithelial ovarian carcinoma (EOC) is related to tumor-specific defects in homologous recombination (HR) and extends beyond BRCA1/2 deficient EOC. A robust method with which to identify HR-deficient (HRD) carcinomas is therefore of utmost clinical importance. In this study, we investigated the proficiency of a functional HR assay based on the detection of RAD51 foci, the REcombination CAPacity (RECAP) test, in identifying HRD tumors in a cohort of prospectively collected epithelial ovarian carcinomas (EOCs). Of the 39 high-grade serous ovarian carcinomas (HGSOC), the RECAP test detected 26% (10/39) to be HRD, whereas ovarian carcinomas of other histologic subtypes (n = 10) were all HR-proficient (HRP). Of the HRD tumors that could be sequenced, 8/9 showed pathogenic BRCA1/2 variants or BRCA1 promoter hypermethylation, indicating that the RECAP test reliably identifies HRD, including but not limited to tumors related to BRCA1/2 deficiency. Furthermore, we found a trend towards better overall survival (OS) of HGSOC patients with RECAP-identified HRD tumors compared to patients with HRP tumors. This study shows that the RECAP test is an attractive alternative to DNA-based HRD tests, and further development of a clinical grade RECAP test is clearly warranted.
Sigma-Aldrich
Freezing container, Nalgene® Mr. Frosty, H × diam. 86 mm × 117 mm
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse