MilliporeSigma
  • The impact of nitrogen oxides on electrochemical carbon dioxide reduction.
The impact of nitrogen oxides on electrochemical carbon dioxide reduction.
Nature communications (2020-11-19)
Byung Hee Ko, Bjorn Hasa, Haeun Shin, Emily Jeng, Sean Overa, Wilson Chen, Feng Jiao
ABSTRACT
The electroreduction of carbon dioxide offers a promising avenue to produce valuable fuels and chemicals using greenhouse gas carbon dioxide as the carbon feedstock. Because industrial carbon dioxide point sources often contain numerous contaminants, such as nitrogen oxides, understanding the potential impact of contaminants on carbon dioxide electrolysis is crucial for practical applications. Herein, we investigate the impact of various nitrogen oxides, including nitric oxide, nitrogen dioxide, and nitrous oxide, on carbon dioxide electroreduction on three model electrocatalysts (i.e., copper, silver, and tin). We demonstrate that the presence of nitrogen oxides (up to 0.83%) in the carbon dioxide feed leads to a considerable Faradaic efficiency loss in carbon dioxide electroreduction, which is caused by the preferential electroreduction of nitrogen oxides over carbon dioxide. The primary products of nitrogen oxides electroreduction include nitrous oxide, nitrogen, hydroxylamine, and ammonia. Despite the loss in Faradaic efficiency, the electrocatalysts exhibit similar carbon dioxide reduction performances once a pure carbon dioxide feed is restored, indicating a negligible long-term impact of nitrogen oxides on the catalytic properties of the model catalysts.
Sigma-Aldrich
Phenol nitroprusside solution
Sigma-Aldrich
Indophenol Blue, Dye content 60 %
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Alkaline Hypochlorite solution