MilliporeSigma
  • The mTOR regulated RNA-binding protein LARP1 requires PABPC1 for guided mRNA interaction.
The mTOR regulated RNA-binding protein LARP1 requires PABPC1 for guided mRNA interaction.
Nucleic acids research (2020-12-18)
Ewan M Smith, Nour El Houda Benbahouche, Katherine Morris, Ania Wilczynska, Sarah Gillen, Tobias Schmidt, Hedda A Meijer, Rebekah Jukes-Jones, Kelvin Cain, Carolyn Jones, Mark Stoneley, Joseph A Waldron, Cameron Bell, Bruno D Fonseca, Sarah Blagden, Anne E Willis, Martin Bushell
ABSTRACT
The mammalian target of rapamycin (mTOR) is a critical regulator of cell growth, integrating multiple signalling cues and pathways. Key among the downstream activities of mTOR is the control of the protein synthesis machinery. This is achieved, in part, via the co-ordinated regulation of mRNAs that contain a terminal oligopyrimidine tract (TOP) at their 5'ends, although the mechanisms by which this occurs downstream of mTOR signalling are still unclear. We used RNA-binding protein (RBP) capture to identify changes in the protein-RNA interaction landscape following mTOR inhibition. Upon mTOR inhibition, the binding of LARP1 to a number of mRNAs, including TOP-containing mRNAs, increased. Importantly, non-TOP-containing mRNAs bound by LARP1 are in a translationally-repressed state, even under control conditions. The mRNA interactome of the LARP1-associated protein PABPC1 was found to have a high degree of overlap with that of LARP1 and our data show that PABPC1 is required for the association of LARP1 with its specific mRNA targets. Finally, we demonstrate that mRNAs, including those encoding proteins critical for cell growth and survival, are translationally repressed when bound by both LARP1 and PABPC1.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human LARP1
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial