MilliporeSigma
  • Refinement of the Peroxidase Peptide Reactivity Assay and Prediction Model for Assessing Skin Sensitization Potential.
Refinement of the Peroxidase Peptide Reactivity Assay and Prediction Model for Assessing Skin Sensitization Potential.
Toxicological sciences : an official journal of the Society of Toxicology (2020-10-24)
Cindy A Ryan, John A Troutman, Petra S Kern, Mike Quijano, Roy L M Dobson, Hong Jian Dai, Thomas M Burt, G Frank Gerberick
ABSTRACT
A peptide reactivity assay with an activation component was developed for use in screening chemicals for skin sensitization potential. A horseradish peroxidase-hydrogen peroxide (HRP/P) oxidation system was incorporated into the assay for characterizing reactivity of hapten and pre-/prohapten sensitizers. The assay, named the Peroxidase Peptide Reactivity Assay (PPRA) had a predictive accuracy of 83% (relative to the local lymph node assay) with the original protocol and prediction model. However, apparent false positives attributed to cysteine depletion at relatively high chemical concentrations and, for some chemicals expected to react with the -NH2 group of lysine, little to no depletion of the lysine peptide were observed. To improve the PPRA, cysteine peptide reactions with and without HRP/P were modified by increasing the number of test concentrations and refining their range. In addition, removal of DL-dithiothreitol from the reaction without HRP/P increased cysteine depletion and improved detection of reactive aldehydes and thiazolines without compromising the assay's ability to detect prohaptens. Modification of the lysine reaction mixture by changing the buffer from 0.1 M ammonium acetate buffer (pH 10.2) to 0.1 M phosphate buffer (pH 7.4) and increasing the level of organic solvent from 1% to 25% resulted in increased lysine depletion for known lysine reactive chemicals. Refinement of the prediction model improved the sensitivity, specificity, and accuracy for hazard identification. These changes resulted in significant improvement of the PPRA making it is a reliable method for predicting the skin sensitization potential of all chemicals, including pre-/prohaptens and directly reactive haptens.
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Peroxidase from horseradish, Type X, ammonium sulfate suspension
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Deferoxamine mesylate salt, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%