MilliporeSigma
  • In vivo ESR imaging of redox status in mice after X-ray irradiation, measured by acyl-protected hydroxylamine probe, ACP.
In vivo ESR imaging of redox status in mice after X-ray irradiation, measured by acyl-protected hydroxylamine probe, ACP.
Free radical biology & medicine (2020-09-07)
Keita Saito, Shoko Okazaki, Yoko Tachibana, Kazunori Anzai, Toshihiko Ozawa, Keizo Takeshita
ABSTRACT
More detailed investigations on the in vivo redox status are needed to elucidate the mechanisms contributing to damage caused by ionizing radiation. In the present study, the in vivo redox status of mice was examined using in vivo electron spin resonance (ESR) imaging after an intraperitoneal injection of 1-acetoxy-3-carbamoyl-2,2,5,5-tetramethylpyrrolidine (ACP) as a probe. ACP is easily hydrolyzed to its hydroxylamine form in the mouse body, and the interconversion between hydroxylamine and the corresponding nitroxyl radical reflects the biological redox status. Liver damage, based on changes in liver weight and plasma aspartate aminotransferase levels, was detected in mice 4 days after X-ray irradiation at 7.5 Gy. ESR imaging showed that the signal intensity of the nitroxyl radical was high at the liver area in both damaged and healthy mice after administration of ACP. Whereas the signal decayed at the liver area for healthy mouse, the decay was negligible in damaged mice. Unlike healthy mouse, signal in the chest for damaged mouse increased with time. The distribution of the sum of hydroxylamine and the nitroxyl radical was similar in damaged and healthy mice. X-ray irradiation slightly lowered the reduction activity of the liver microsomal fraction for the nitroxyl radical. Thiobarbituric acid reactive substances in the liver were higher in damaged mice than in healthy mice; however, no significant differences were noted in reduced glutathione. The present results indicate that the redox status of mice exposed to X-ray irradiation is more oxidative than that in healthy mice.