MilliporeSigma
  • Temporal trends of "old" and "new" persistent halogenated organic pollutants in fish from the third largest freshwater lake in China during 2011-2018 and the associated health risks.
Temporal trends of "old" and "new" persistent halogenated organic pollutants in fish from the third largest freshwater lake in China during 2011-2018 and the associated health risks.
Environmental pollution (Barking, Essex : 1987) (2020-09-06)
Jinjing Ma, Xiangnan Li, Shengtao Ma, Xiaolan Zhang, Guiying Li, Yingxin Yu
ABSTRACT
The study aimed to investigate temporal trends of "old" and "new" persistent halogenated organic pollutants (HOPs) in Taihu Lake, the third largest freshwater lake in China, and the associated health risks. Five fish species were consecutively collected from the lake every year during 2011-2018. HOPs including 37 polychlorinated biphenyls (PCBs), 10 organochlorine pesticides (OCPs), short- and medium-chain chlorinated paraffins (SCCPs and MCCPs), 19 polybrominated diphenyl ethers (PBDEs), and 10 new brominated flame retardants (NBFRs), were measured. The results showed that all the HOPs were detected, with MCCPs and NBFRs showing the highest and lowest concentrations, respectively. The levels of SCCPs and MCCPs were several orders of magnitude higher than those of the other HOPs. There were obvious increasing trends for SCCPs, MCCPs, and hexachlorobenzene, but a decreasing trend for PBDEs. No obvious increasing or decreasing trends were observed for the other HOPs. The present study indicated that the use of NBFRs to replace PBDEs was not yet clearly observed. Fish consumption did not result in non-carcinogenic risks, but posed low carcinogenic risks, with PCBs and DDTs being the highest-risk contaminants because of historical residues. This is the first study for the temporal variations of the HOPs in the lake.
Sigma-Aldrich
2,4,4,6-Tetrabromo-2,5-cyclohexadienone, 90%