MilliporeSigma
  • Identification of a Male-Produced Volatile Pheromone for Phymatodes dimidiatus (Coleoptera: Cerambycidae) and Seasonal Flight Phenology of Four Phymatodes Species Endemic to the North American Intermountain West.
Identification of a Male-Produced Volatile Pheromone for Phymatodes dimidiatus (Coleoptera: Cerambycidae) and Seasonal Flight Phenology of Four Phymatodes Species Endemic to the North American Intermountain West.
Environmental entomology (2020-09-05)
Claudia D Lyons-Yerion, James D Barbour, Judith A Mongold-Diers, Christopher J Williams, Stephen P Cook
ABSTRACT
Research over the last 15 yr has shown widespread pheromone parsimony within the coleopteran family Cerambycidae, with a number of highly conserved pheromone motifs, often shared within and across subfamilies, tribes, and genera. Our goals were to increase our understanding of the evolution of volatile pheromones within the Cerambycidae, their role in reproductive isolation and to identify pheromones for use in the development of lures for monitoring cerambycids. Over 3 yr, we tested 12 compounds known to be cerambycid pheromones as possible attractants at sites across Idaho. This study focused on species within the cerambycine genus Phymatodes (Tribe: Callidiini). We also collected and analyzed headspace volatiles of captured Phymatodes dimidiatus (Kirby). Our results demonstrate that (R)-2-methylbutan-1-ol is a male-produced volatile pheromone for P. dimidiatus. These results are consistent with prior research suggesting that (R)-2-methylbutan-1-ol and (R)-3-hydroxyhexan-2-one, individually or in a blend of both compounds, commonly serve as pheromones for Phymatodes spp. We captured Phymatodes starting in mid-May, continuing through mid-August. Our data indicate that flight periods of Phymatodes spp. in Idaho overlap. These species may be utilizing various mechanisms to ensure reproductive isolation, such as the production of different volatile pheromones, minor components, and/or proportions of components, utilizing different host species and/or host volatiles, differing daily activity periods, and/or occupying different heights in the tree canopy. Our results contribute to the basic understanding of the chemical and behavioral ecology of the Cerambycidae and can be applied to the development of pheromone lures for monitoring of economically important or endangered species.
Sigma-Aldrich
2-(undecyloxy)ethanol, AldrichCPR