MilliporeSigma
  • Constitutively active HCN channels constrain detrusor excitability and modulate evoked contractions of human bladder.
Constitutively active HCN channels constrain detrusor excitability and modulate evoked contractions of human bladder.
American journal of clinical and experimental urology (2020-11-26)
Mahendra Kashyap, Nishant Singh, Naoki Yoshimura, Christopher Chermansky, Pradeep Tyagi
ABSTRACT
Expression of Hyperpolarization-activated cyclic nucleotide-gated (HCN) channels is reported in bladder, but the functional role remains unsettled. Here, we immunolocalized the HCN1 and HCN4 subtype in human bladder and investigated their functional significance. Bladder procured from ten organ donors was dissected into mucosa (containing urothelium and submucosa) and detrusor for double immunofluorescence of HCN1 and 4 subtypes with gap junction and neural proteins together with isometric tension recordings. Mucosa intact and denuded detrusor strips were stretched to a basal tension of 10 mN for eliciting either tetrodotoxin (TTX) resistant spontaneous, carbachol evoked contractions and TTX sensitive electrical field stimulated (EFS), pre and post-addition of HCN blocker, ZD7288 or the activator, Lamotrigine or the cholinesterase inhibitor, Neostigmine. Double immunofluorescence revealed immunolocalization of HCN1 and HCN4 subtype with calcitonin gene related peptide (CGRP), choline acetyl transferase and gap junction proteins in mucosa and detrusor. Removal of mucosa significantly raised the resting tension and the force of spontaneous contractions upon cumulative addition of ZD7288 in micromolar range relative to Lamotrigine treated strips (P<0.05). ZD7288 [10 nM] did not affect the contractile response evoked by EFS or carbachol, but the addition of ZD7288 [10 nM] in presence of Neostigmine [1 µM] significantly enhanced the atropine and TTX sensitive EFS evoked contractions of mucosa denuded strips. Overall, HCN channels immunolocalized in mucosa, smooth muscle, gap junctions and nerve fibers exert a tonic constraint on detrusor excitability, enable spatio-temporal integration of evoked contractions and constrain the release of neurotransmitters, respectively. In contrast to the pacemaker role in other organs, findings argue for a non-pacemaking role of HCN channels in human bladder.
Sigma-Aldrich
Anti-Connexin 45 Antibody, near CT, cytoplasmic, clone 8A11.2, clone 8A11.2, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Pricing and availability is not currently available.