MilliporeSigma
  • Aberrant ER Stress Induced Neuronal-IFNβ Elicits White Matter Injury Due to Microglial Activation and T-Cell Infiltration after TBI.
Aberrant ER Stress Induced Neuronal-IFNβ Elicits White Matter Injury Due to Microglial Activation and T-Cell Infiltration after TBI.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2019-11-07)
Tanusree Sen, Pampa Saha, Rajaneesh Gupta, Lesley M Foley, Tong Jiang, Olena S Abakumova, T Kevin Hitchens, Nilkantha Sen
ABSTRACT
Persistent endoplasmic reticulum (ER) stress in neurons is associated with activation of inflammatory cells and subsequent neuroinflammation following traumatic brain injury (TBI); however, the underlying mechanism remains elusive. We found that induction of neuronal-ER stress, which was mostly characterized by an increase in phosphorylation of a protein kinase R-like ER kinase (PERK) leads to release of excess interferon (IFN)β due to atypical activation of the neuronal-STING signaling pathway. IFNβ enforced activation and polarization of the primary microglial cells to inflammatory M1 phenotype with the secretion of a proinflammatory chemokine CXCL10 due to activation of STAT1 signaling. The secreted CXCL10, in turn, stimulated the T-cell infiltration by serving as the ligand and chemoattractant for CXCR3+ T-helper 1 (Th1) cells. The activation of microglial cells and infiltration of Th1 cells resulted in white matter injury, characterized by impaired myelin basic protein and neurofilament NF200, the reduced thickness of corpus callosum and external capsule, and decline of mature oligodendrocytes and oligodendrocyte precursor cells. Intranasal delivery of CXCL10 siRNA blocked Th1 infiltration but did not fully rescue microglial activation and white matter injury after TBI. However, impeding PERK-phosphorylation through the administration of GSK2656157 abrogated neuronal induction of IFNβ, switched microglial polarization to M2 phenotype, prevented Th1 infiltration, and increased Th2 and Treg levels. These events ultimately attenuated the white matter injury and improved anxiety and depressive-like behavior following TBI.SIGNIFICANCE STATEMENT A recent clinical study showed that human brain trauma patients had enhanced expression of type-1 IFN; suggests that type-1 IFN signaling may potentially influence clinical outcome in TBI patients. However, it was not understood how TBI leads to an increase in IFNβ and whether induction of IFNβ has any influence on neuroinflammation, which is the primary reason for morbidity and mortality in TBI. Our study suggests that induction of PERK phosphorylation, a characteristic feature of ER stress is responsible for an increase in neuronal IFNβ, which, in turn, activates microglial cells and subsequently manifests the infiltration of T cells to induce neuroinflammation and subsequently white matter injury. Blocking PERK phosphorylation using GSK2656157 (or PERK knockdown) the whole cascade of neuroinflammation was attenuated and improved cognitive function after TBI.
Sigma-Aldrich
Anti-STING Antibody, clone S17G2C4B7, clone S17G2C4B7, from mouse
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody, serum, from guinea pig